Human recombinant LAIR2 (from cells)
Fournisseur: ProSci Inc.
91-340
PRSI91-340EA 549 EUR InStock
PRSI91-340
Human recombinant LAIR2 (from cells)
Protéines et Peptides
Leukocyte-Associated Immunoglobulin-Like Receptor 2 (LAIR2) is a secreted, 131 amino acid protein that contains one Ig-like C2 type domain, making it a member of the Ig superfamily. When compared to LAIR-1, its transmembrane counterpart, it shares 83% amino acid identity across the signal sequence and extracellular domains; although one is secreted and one is membrane-bound, the two LAIR proteins are thought to have arisen from a common gene ancestor and appear to share similar adhesion profiles. This suggests that LAIR-2 may compete with LAIR-1 for ligand binding. A 114 amino acid alternate splice form of LAIR-2 is truncated at the C terminus, but retains the entire Ig domain. The expression profile of these splice forms, and the presence of orthologs in other species, have not been reported.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité