Human recombinant LBP (from cells)
Fournisseur: ProSci Inc.
91-341
PRSI91-341EA 734 EUR InStock
PRSI91-341
Human recombinant LBP (from cells)
Protéines et Peptides
Lipopolysaccharide binding protein (LBP) is a plasma protein, belongs to a member of structurally and functionally related proteins which includes bactericidal permeability-increasing protein (BPI), plasma cholesteryl ester transfer protein (CETP) and phospholipid transfer protein (PLTP). It is involved in the acute-phase immunologic response to gram-negative bacterial infections. In cooperation with BPI. LBP binds LPS and interacts with the CD14 receptor, most likely playing a role in regulating LPS-dependent monocyte responses. Studies suggest that LBP is necessary for the rapid acute-phase response to LPS but not for the clearance of LPS from circulation. Finally, t The LBP gene is found on chromosome 20, directly downstream of the BPI gene.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité