Human recombinant Legumain (from cells)
Fournisseur: ProSci Inc.
91-342
PRSI91-342EA 884 EUR InStock
PRSI91-342
Human recombinant Legumain (from cells)
Protéines et Peptides
Legumain is a lysosomal cysteine protease which is a member of the peptidase C13 family. Though it is found in many tissues, it is highly expressed in the kidney, heart, and placenta. Legumain has a strict specificity for hydrolysis of asparaginyl bonds and can also cleave aspartyl bonds slowly, especially under acidic conditions. Over-expression Legumain in tumors is significant for invasion and metastasis. In addition, Legumain may be involved in the processing of proteins for MHC class II antigen presentation in the lysosomal/endosomal system and negative regulation of neuron apoptosis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité