Human recombinant lumican (from cells)
Fournisseur: ProSci Inc.
91-343
PRSI91-343EA 549 EUR InStock
PRSI91-343
Human recombinant lumican (from cells)
Protéines et Peptides
Lumican is a 40 kD secreted protein which belongs to the small leucine-rich repeat proteoglycans (SLRPs) and the class II subfamily. Human Lumican is synthesized as a 338 amino acid precursor then cut the 18 aa signal sequence. The mature Human Lumican contains 12 leucine-rich repeats (LRRs), 4 potential sites of N-linked glycosylation, and a C- terminal with two conserved cyst-eines. Lumican can be existed in extracellular matrix of human articular cartilage. Lumican participates in the maintenance of tissue homeostasis and regulates cellular functions in vivo, such as cell proliferation, adhesion, migration, and differentiation. The overexpression of lumican has been correlated to colorectal tumor, breast, neuroendocrine, and pancreatic cancers.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité