Human recombinant MMP2 (from cells)
Fournisseur: ProSci Inc.
91-345
PRSI91-345EA 852 EUR InStock
PRSI91-345
Human recombinant MMP2 (from cells)
Protéines et Peptides
72 kDa type IV collagenase also known as matrix metalloproteinase-2 (MMP-2) and gelatinase A is an enzyme that in humans is encoded by the MMP2 gene.It belongs to the matrix metalloproteinase (MMP) family. Matrix metalloproteinases (MMPs) are a family of zinc-dependent endopeptidases that degrade components of the extracellular matrix (ECM) and play essential roles in various physiological processes such as morphogenesis, differentiation, angiogenesis and tissue remodeling, as well as pathological processes including inflammation, arthritis, cardiovascular diseases, pulmonary diseases and tumor invasion. MMP-2 is ubiquitinous metalloproteinase that is involved in diverse functions such as remodeling of the vasculature, angiogenesis, tissue repair, tumor invasion, inflammation, atherosclerotic plaque rupture, as well as degrading extracellular matrix proteins. MMP-2 can also act on several nonmatrix proteins such as big endothelial 1 and beta-type CGRP promoting vasoconstriction. MMP-2 cleaves KISS at a Gly-|-Leu bond and appears to have a role in myocardial cell death pathways.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité