Human recombinant poly-Ig receptor (from cells)
Fournisseur: ProSci Inc.
91-349
PRSI91-349EA 549 EUR InStock
PRSI91-349
Human recombinant poly-Ig receptor (from cells)
Protéines et Peptides
The human Polymeric Immunoglobulin Receptor (pIgR) is a 100 kDa type I transmembrane glycoprotein. Its precursor is 764 amino acids. It contains an 18 amino acid signal sequence, a 620 amino acid extracellular region, a 23 amino acid transmembrane fragment, and a 103 amino acid cytoplasmic domain. pIgR is synthesized by secretory epithelial cells with five Ig-like domains in extracellular region, and transfer to the basolateral plasma membrane. For IgA and IgM polymers, in addition to alpha-heavy chains and light Ig chains, a short polypeptide named joining chain (J chain) is also contained and required. pIgR can bind larger polymers of IgA (pIgA) and pentameric IgM as a carrier that transports IgA and IgM across epithelium. The receptor-ligand complexes are endocytosed and transcytosed to the apical surface, then proteolytic cleavage of the sixth extracellular domain of pIgR and generate secretory IgA (SIgA), the pIgR fragment is referred to as secretory component (SC). SIgA is a important component of the mucosal immune system. SC is anti-microbial properties and protects SIgA from proteolytic degradation
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité