Human recombinant Biglycan (from Cells)
Fournisseur: ProSci Inc.
91-353
PRSI91-353EA 654 EUR InStock
PRSI91-353
Human recombinant Biglycan (from Cells)
Protéines et Peptides
Biglycan is a 200-350 kD proteoglycan consisting of a 45 kD core protein and two chrondroitin/dermatan sulfate glycosaminoglycan chains. Biglycan binds to TGF-ß. It also binds to collagen type I in low ionic strength (less than 3 mM phosphate) buffer. At higher ionic strengths, Biglycan does not bind to collagen type I. It enhances the inhibition effect of TGF-ß on osteoclast proliferation at a concentration of 4-20 mg/ml. It also prevents the attachment of CHO cells to fibronectin, with a 50% inhibition at 17-21 mg/ml.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité