Human recombinant SPARC (from cells)
Fournisseur: ProSci Inc.
91-355
PRSI91-355EA 409 EUR InStock
PRSI91-355
Human recombinant SPARC (from cells)
Protéines et Peptides
Secreted Protein Acidic and Rich in Cysteine (SPARC) is a secreted, evolutionarily conserved collagen-binding glycoprotein and belongs to the SPARC family. SPARC has 286 amino acids and contains an EF-hand in C-termina domain, a follistatin-like domain with Kazal-like sequences. There are two calcium binding sites, one binds 5 - 8 Ca2+ with a low affinity and other on an EF-hand loop that binds a Ca2+ ion with a high affinity. It is highly expressed in tissues undergoing morphogenesis, remodeling and wound repair. SPARC regulate cell growth through interactions with the extracellular matrix (ECM) and cytokines. SPARC bind to numerous proteins of the ECM, affect ECM protein expression, influence cellular adhesion and migration, and modulate growth factor-induced cell proliferation and angiogenesis. SPARC also binds several types of collagen, albumin, thrombospondin, PDGF and cell membranes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité