Human recombinant TWSG1 (from cells)
Fournisseur: ProSci Inc.
91-359
PRSI91-359EA 734 EUR InStock
PRSI91-359
Human recombinant TWSG1 (from cells)
Protéines et Peptides
Twisted Gastrulation Protein Homolog 1 (TWSG1) is a 22 kDa secreted protein that belongs to the twisted gastrulation protein family. Human TWSG1 is synthesized as a 223 aa precursor that contains a 25 aa signal peptide and a 198 aa mature chain. TWSG1 may be involved in dorsoventral axis formation. TWSG1 seems to antagonize BMP signaling by forming ternary complexes with CHRD and BMPs, thereby preventing BMPs from binding to their receptors.TWSG1 can inhibit BMP activity by binding directly to BMP proteins, and can act the anti-BMP function, partly mediated by cleavage and degradation of CHRD, which releases BMPs from ternary complexes. TWSG1 may be an important modulator of BMP-regulated cartilage development, chondrocyte differentiation and thymocyte development.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité