Human recombinant UPA (from cells)
Fournisseur: ProSci Inc.
91-360
PRSI91-360EA 884 EUR InStock
PRSI91-360
Human recombinant UPA (from cells)
Protéines et Peptides
Recombinant Human Urokinase-Type Plasminogen Activator is a serine protease, which specifically cleaves the zymogen plasminogen to form the active enzyme plasmin. Urokinase-Type Plasminogen Activator is a potent marker of invasion and metastasis in many human cancers associated with breast, colon, stomach, bladder, brain, ovary and endometrium. Human Urokinase-Type Plasminogen Activator is initially synthesized as 431 amino acid precursor with a N-terminal signal peptide residues. The single chain molecule is processed into a disulfide-linked two-chain molecule. There exists two forms A chain, the long A chain contains an EGF-like domain that is responsible for binding of the uPA receptor. The B chain corresponds to the catalytic domain.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité