Human recombinant Dickkopf-Related 3 (from cells)
Fournisseur: ProSci Inc.
91-373
PRSI91-373EA 549 EUR InStock
PRSI91-373
Human recombinant Dickkopf-Related 3 (from cells)
Protéines et Peptides
Dickkopf-related protein 3 (DKK3) belongs to the DKK protein family including Dkk-1, 2, 3 and -4. DKK3 is a 350 amino acid secreted glycoprotein which is comprised of an N-terminal signal peptide and 2 conserved cysteine-rich domains that are separated by a 12 amino acid linker region. Dkk-3 also have one prokineticin domain. DKK3 is involved in embryonic development through its inhibition of the WNT signaling pathway. The Dkk family also includes Soggy, which is homologous to Dkk-3 but not to the other family members. Soggy has not been shown to inhibit Wnt signaling, and its role in the pathway is unclear.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité