Human recombinant SP-10 (from cells)
Fournisseur: ProSci Inc.
91-378
PRSI91-378EA 654 EUR InStock
PRSI91-378
Human recombinant SP-10 (from cells)
Protéines et Peptides
Acrosomal Protein SP-10 is a testis-specific differentiation antigen that is associated with acrosomal membranes and matrix of mature sperm. It has been detected in several species including humans. Acrosomal Protein SP-10 may be involved in sperm-zona binding or penetration as a potential contraceptive vaccine immunogen for humans. ACRV1 is also a intra-acrosomal protein that is considered to be a vaccine candidate for immunocontraception.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité