Human recombinant ASAM (from Cells)
Fournisseur: ProSci Inc.
91-383
PRSI91-383EA 549 EUR InStock
PRSI91-383
Human recombinant ASAM (from Cells)
Protéines et Peptides
Adipocyte Adhesion Molecule (ASAM) is a type I transmembrane protein and member of the CTX family within the immunoglobulin superfamily. ASAM may be involved in the cell-cell adhesion, play an important role in adipocyte differentiation and development of obesity. ASAM can be expressed in the skeletal, heart, colon, spleen, muscle, lung and kidney with high level, and in the peripheral blood leukocytes and liver with low level. The extracellular region of ASAM consists two potential N-linked glycosylation sites, and two immunoglobulin domains, one V-type and one C2-type.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité