Human recombinant EMMPRIN (from cells)
Fournisseur: ProSci Inc.
91-389
PRSI91-389EA 588 EUR InStock
PRSI91-389
Human recombinant EMMPRIN (from cells)
Protéines et Peptides
Extracellular Matrix Metalloproteinase Inducer (EMMPRIN) belongs to the immunoglobulin superfamily, which has the homology to both the immunoglobulin V domain and MHC class II antigen beta -chain. EMMPRIN is a transmembrane glycoprotein with different forms, resulting from different modes of glycosylation and N-terminal sequence variants. EMMPRIN can be expressed in breast cancer, oral squamous cell carcinoma, glioma, lymphoma, lung, bladder, and melanoma carcinomas cells. EMMPRIN promotes invasion, metastasis, growth, and survival of malignants cells, serves as a receptor for extracellular cyclophilinthe, may play a role in signal transduction.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité