Human recombinant CADM3 (from Cells)
Fournisseur: ProSci Inc.
91-391
PRSI91-391EA 471 EUR InStock
PRSI91-391
Human recombinant CADM3 (from Cells)
Protéines et Peptides
Cell Adhesion Molecular Proteins are proteins located on the cell surface involved with the binding with other cells or with the extracellular matrix in the cell adhesion process. These proteins consists of three domains, an transmembrane domain, an intracellular domain that interacts with the cytoskeleton, and an extracellular domain that interacts with other CAMs of the same kind or with other CAMs or the extracellular matrix. Cell Adhesion Molecular 3 (CADM3) is a neural tissue-specific member of the nectin-like family of immunoglobulin superfamily. CADM3 interacts with EPB41L1 may regulate structure or function of cell-cell junctions. CADM3 has both calcium-independent homophilic cell-cell adhesion activity and calcium-independent heterophilic cell-cell adhesion activity with IGSF4, PVRL1 and PVRL3.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité