Human recombinant CXCL9 (from cells)
Fournisseur: ProSci Inc.
91-393
PRSI91-393EA 734 EUR InStock
PRSI91-393
Human recombinant CXCL9 (from cells)
Protéines et Peptides
Chemokine (C-X-C Motif) Ligand 9 (CXCL9) belongs to the intercrine alpha (chemokine CXC) family. It is secreted by interferon stimulated monocytes, macrophages and endothelial cells, which elicits chemotactic functions by interacting with the chemokine receptor CXCR3. CXCL9 acts as a Th1 (type 1 helper T) cell chemoattractant and plays a role in the growth, activation and movement of cells associated with immune and inflammatory responses, and in tumour growth inhibition. It is closely related to two other CXC chemokines called CXCL10 and CXCL11, whose genes are located near the gene for CXCL9 on human chromosome 4.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité