Human recombinant Cathepsin Z (from Cells)
Fournisseur: ProSci Inc.
91-394
PRSI91-394EA 789 EUR InStock
PRSI91-394
Human recombinant Cathepsin Z (from Cells)
Protéines et Peptides
Cathepsin Z is a lysosomal cysteine proteinase and belongs to the peptidase C1 family. Human Cathepsin Z contains a singnal sequence, a propeptide and a mature chain. It exhibits bothcarboxy-monopeptidase and carboxy-dipeptidase activities. In contrast to cathepsin B, it does not act as an endopeptidase. Cathepsin Z is restricted to the cells of theimmune system, predominantly monocytes, macrophages and dendritic cells. It is expressed ubiquitously in cancer cell lines and primary tumors. Like other members of this family, Cathepsin Z may be involved in tumorigenesis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité