Human recombinant Fc gamma RIIIA (from cells)
Fournisseur: ProSci Inc.
91-397
PRSI91-397EA 490 EUR InStock
PRSI91-397
Human recombinant Fc gamma RIIIA (from cells)
Protéines et Peptides
Receptors for the Fc region of immunoglobin G (Fc gamma R) are divided into three classes and Fc gamma RIII is a multifunctional, low/intermediate affinity receptor. In humans, Fc gamma RIII is expressed as two distinct forms (Fc gamma RIIIA and Fc gamma RIIIB) that are encoded by two different but highly homologous genes in a cell type-specific manner. Fc gamma RIIIB is a low-affinity, GPI-linked receptor expressed by neutrophils and eosinophils, whereas Fc gamma RIIIA is an intermediate affinity polypeptide-anchored transmembrane glycoprotein expressed by a subset of T lymphocytes, natural killer (NK) cells, monocytes, and macrophages. The Fc gamma RIIIA receptor is involved in phagocytosis, secretion of enzymes, inflammatory mediators, antibody-dependent cellular cytotoxicity (ADCC), mast cell degranulation, and clearance of immune complexes. Fc gamma RIIIA has an immunoreceptor tyrosine-based activation motif (ITAM) in its cytoplasmic domain and delivers an activation signal in the immune responses. Aberrant expression or mutations in this gene is implicated in susceptibility to recurrent viral infections, systemic lupus erythematosus, and alloimmune neonatal neutropenia. In humans, it is a 50 -70 kD type I transmembrane activating receptor. The Fc gamma RIIIA cDNA encodes 254 amino acid including a 16aa signal sequence, 191 amino acid ECD with two C2-type Ig-like domains, five potential N-glycosylation sites, a 22 amino acid transmembrane sequence and a 25 amino acid cytoplasmic domain.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité