Human recombinant siglec-3 (from cells)
Fournisseur: ProSci Inc.
91-401
PRSI91-401EA 490 EUR InStock
PRSI91-401
Human recombinant siglec-3 (from cells)
Protéines et Peptides
CD33 is a type I Lectin belonging to the Ig superfamily. CD33 contains an N terminal Ig like V type domain, which mediates sialic acid binding, followed by one Ig like C2 type domain, a transmembrane region and a cytoplasmic tail containing two conserved immunoreceptor tyrosine based inhibition motifs (ITIMs). Eleven human Siglecs have been characterized. Siglecs 5 to 11 share a high degree of sequence similarity with CD33/Siglec3 both in their extracellular and intracellular regions. They are collectively referred to as CD33 related Siglecs. CD33 related Siglecs have differential expression pattern within the hematopoietic system. They are involved in the regulation of cellular activation within the immune system. Siglec 3 expression is restricted to cells of myelomonocytic lineage. Siglec3 recruits SHP1 and SHP2 to its ITIMs upon phosphorylation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité