Human recombinant 5'-Nucleotidase (from cells)
Fournisseur: ProSci Inc.
91-402
PRSI91-402EA 789 EUR InStock
PRSI91-402
Human recombinant 5'-Nucleotidase (from cells)
Enzymes
CD73 is a glycosyl phosphatidylinositol (GPI) anchored membrane protein that belongs to the 5'-nucleotidase family. CD73 is an ecto 5'Nucleotidase expressed by most cell types. CD73 hydrolyses extracellular nucleotides into membrane permeable nucleosides. CD73 is one of several enzymes responsible for the production of extracellular adenosine, a signaling molecule that is involved in responses to inflammation and tissue injury. CD73 is a lymphocyte maturation marker that has functions independent of its catalytic activity. CD73 is also a regulator of leukocyte extravasation, a function that requires its 5'Nucleotidase activity. Defects in NT5E are the cause of calcification of joints and arteries (CAJA). The recombinant CD73 lacking GPI anchor is secreted as a monomer.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité