L'expérience en ligne d'Avantor évolue pour mieux vous accompagner ! À compter du 4 avril 2025, nos clients seront migrés vers une nouvelle plateforme pour une expérience d'achat en ligne simplifiée.
Human recombinant Carboxylesterase 1 (from Cells)
Fournisseur: ProSci Inc.
91-406
PRSI91-406EA 884 EUR InStock
PRSI91-406
Human recombinant Carboxylesterase 1 (from Cells)
Enzymes
Carboxylesterase 1 (CES1) is a member of a large family of carboxylesterases that are responsible for the hydrolysis of ester and amide bonds. These enzymes have broad substrate specificity ranging from small molecule esters such as phenylester to long chain fatty acid esters and thioesters. They are major determinants of the pharmacokinetic behavior of most therapeutic agents containing an ester or amide bond. CES1 shares the serine hydrolase fold observed in other esterases. CES1 hydrolyses aromatic and aliphatic esters, but has no catalytic activity toward amides or a fatty acyl-CoA ester. CES1 participates in detoxification of drugs such as cocaine and heroin in serum and liver. It may also play a role in detoxification in the lung and/or protection of the central nervous system from ester or amide compounds.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité