Human recombinant Chitinase-3-like 1 (from cells)
Fournisseur: ProSci Inc.
91-408
PRSI91-408EA 549 EUR InStock
PRSI91-408
Human recombinant Chitinase-3-like 1 (from cells)
Enzymes
Chitinase-3-Like Protein 1 (CHI3L1) belongs to the glycosyl hydrolase 18 family. CHI3L1 is expressed in activated macrophages, articular chondrocytes, synovial cells as well as in liver. It lacks chitinase activity and is secreted by activated macrophages, chondrocytes, neutrophils and synovial cells. CHI3L1 is thought to play a role in defense against pathogens, or in tissue remodeling, and in the capacity of cells to respond to and cope with changes in their environment. In addition, CHI3L1 is associated with susceptibility to asthma-related traits type 7 (ASRT7) which assessed by methacholine challenge test, serum IgE levels, atopy, and atopic dermatitis.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité