Human recombinant CLEC3B (from cells)
Fournisseur: ProSci Inc.
91-409
PRSI91-409EA 549 EUR InStock
PRSI91-409
Human recombinant CLEC3B (from cells)
Protéines et Peptides
C-Type Lectin Domain Family 3 Member B (CLEC3B) is a serum and tissue protein and it contais a C-type lectin which binds to Ca++. CLEC3B is originally found in plasma, the concentrations approximately 10mg/l. It can bind to kringle 4 of plasminogen and enhance the activation of plaminogen to plasmin, catalyzed by tissue plasminogen activator in the presence of poly-D-lysine. In addition, CLEC3B may be involved in the packaging of molecules destined for exocytosis. Also, CLEC3B is best known as a prognostic marker in ovarian cancer.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité