Human recombinant Cathepsin L2 (from Cells)
Fournisseur: ProSci Inc.
91-416
PRSI91-416EA 789 EUR InStock
PRSI91-416
Human recombinant Cathepsin L2 (from Cells)
Protéines et Peptides
Cathepsin L2 belongs to the Peptidase C1 family. Cathepsin L2 can be autocatalytically convertedto the mature form as a proenzyme in lysosomes at pH=7.0. Cathepsin L2 cannot be expressed in normal mammary gland, colon and peritumoral tissue, but it can be expressed in breast carcinomas and colorectal tissues. These suggeats Cathepsin L2 may play a role in tumor processes. Cathepsin L2 is specifically expressed in the testis, thymus, and corneal epithelium.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité