Human recombinant Ephrin A3 (from cells)
Fournisseur: ProSci Inc.
91-420
PRSI91-420EA 343 EUR InStock
PRSI91-420
Human recombinant Ephrin A3 (from cells)
Protéines et Peptides
Ephrins-A3 belongs the Ephrins ligand family which involved in a variety of biological processes, especially in the nervous system and in erythropoiesis. It is shown that Ephrin-A3 is expressed in brain, skeletal muscle, spleen, thymus, prostate, testis, ovary, small intestine, and peripheral blood leukocytes. Ephrin-A3 has a GPI anchor following the extracellular sequence and a signal sequence of 22 amino acids. Ephrin-A3 can bind EphA2, EphA3, EphA4, EphA5, EphA6, EphA7, EphA8 and EphB1. Futhermore, it is associated with tumor growth and metastasis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité