Human recombinant Ephrin B2 (from cells)
Fournisseur: ProSci Inc.
91-421
PRSI91-421EA 306 EUR InStock
PRSI91-421
Human recombinant Ephrin B2 (from cells)
Protéines et Peptides
Ephrin-B2 is a type I transmembrane protein and belongs the Ephrin family. It binds to the receptor tyrosine kinases, such as EPHA4, EPHB4 and EPHA3. Ephrin-B2 has been implicated in mediating developmental events, especially in the nervous system, erythropoiesis and tumour metastasis. Ligation of Ephrin-B2 with complementary EphB receptors on adjacent cells results in a combination of forward (EphB receptors) and reverse (Ephrin-B2) signalling, which is central to tissue development and remodelling functions. In addition, Ephrin-B2 may play a role in constraining the orientation of longitudinally projecting axons.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité