Human recombinant ERMAP (from cells)
Fournisseur: ProSci Inc.
91-422
PRSI91-422EA 584 EUR InStock
PRSI91-422
Human recombinant ERMAP (from cells)
Protéines et Peptides
Human Erythroid Membrane-Associated Protein (ERMAP) is a cell surface transmembrane protein that belongs to the immunoglobulin superfamily. It is hghly expressed in bone marrow and to a lower extent in leukocytes, thymus, lymph node and spleen. ERMAP contains 1 B30.2/SPRY domain and 1 Ig-like V-type (immunoglobulin-like) domain. It may serve as an erythroid cell receptor, possibly as a mediator of cell adhesion. ERMAP is responsible for the Scianna/Radin blood group system. Two transcript variants encoding the same protein have been found for this gene ERMAP.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité