Human recombinant CD89 (from cells)
Fournisseur: ProSci Inc.
91-424
PRSI91-424EA 549 EUR InStock
PRSI91-424
Human recombinant CD89 (from cells)
Protéines et Peptides
Immunoglobulin alpha Fc Receptor (IgA Fc Receptor) is a member of the immunoglobulin gene superfamily. It is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils, monocytes, macrophages, and eosinophils, where it mediates immunologic responses to pathogens through the charged arginin residue within its transmembrane domain. IgA Fc Receptor binds both IgA1 and IgA2 with similar affinity. The site of interaction between FCAR and IgA was identified in the first extracellular domain of FCAR and the C2/C3 junction of IgA. It interacts with IgA-opsonized targets and triggers several immunologic defense processes, including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and stimulation of the release of inflammatory mediators. FCAR is also expressed on Kupffer cells in the liver, where it was suggested to provide a second line of defense.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité