Rat Recombinant MCSF
Fournisseur: ProSci Inc.
91-425
PRSI91-425EA 761 EUR InStock
PRSI91-425
Rat Recombinant MCSF
Protéines et Peptides
Rat Macrophage colony-stimulating factor 1(MCSF,CSF1) is a single-pass type I membrane cytokine. It is a hematopoietic growth factor that plays an essential role in the regulation of survival, proliferation and differentiation of hematopoietic precursor cells, especially mononuclear phagocytes, such as macrophages and monocytes. MCSF promotes the release of proinflammatory chemokines, and thereby plays an important role in innate immunity and in inflammatory processes. It is involved in the regulation of osteoclast proliferation and differentiation, the regulation of bone resorption, and is required for normal bone development which for normal male and female fertility. It promotes reorganization of the actin cytoskeleton, regulates formation of membrane ruffles, cell adhesion and cell migration. MCSF also plays a role in lipoprotein clearance.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité