Human Recombinant GFR-alpha-2 (from Cells)
Fournisseur: ProSci Inc.
91-427
PRSI91-427EA 377 EUR InStock
PRSI91-427
Human Recombinant GFR-alpha-2 (from Cells)
Protéines et Peptides
Members of the glial cell line-derived neurotrophic factor (GDNF) family, including GDNF and Neurturin, play key roles in the control of vertebrate neuronal survivial and differentiation. GDNF is a glycosylated, disulfide-bonded homodimer that is distantly related to the TGF superfamily of growth factors. Three receptors for these factors, GFR alpha-1, GFR alpha-2, and GFR alpha-3 have been identified. The receptors do not contain transmembrane domains and are attached to the cell membrane by glycosyl-phosphoinositol linkage. Both GFR alpha-1 and GFR alpha-2 have been shown to mediate the GDNF-dependent and Neurturin-dependent phosphorylation and activation of the tyrosine kinase Ret. GFR-3 is expressed only during development. GFR alpha-2 binds Neurturin and mediates activation of RET receptor tyrosine kinase by both Neurturin and GDNF.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité