Human recombinant beta-Galactosidase (from Cells)
Fournisseur: ProSci Inc.
91-428
PRSI91-428EA 734 EUR InStock
PRSI91-428
Human recombinant beta-Galactosidase (from Cells)
Enzymes
beta Galactosidase is a lysosomal beta Galactosidase that hydrolyses the terminal beta Galactose from Ganglioside and Keratan sulfate. In lysosome, the mature beta Galactosidase protein associates with Cathepsin A and Neuraminidase 1 to form the lysosomal multienzyme complex . An alternative splicing at the RNA level of beta Galactosidase results a catalytically inactive beta Galactosidase that plays an important role in vascular development. Defects of beta -galactosidase (GLB1) are the cause of diseases like GM1-gangliosidosis which is a lysosomal storage disease and Morquio Syndrome B that cause patients to have abnormal elastic fibers. More than 100 mutations have been identified for beta Galactosidase, which result in different residual activities of the mutant enzymes and a spectrum of symptoms in the two related diseases.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité