Human recombinant ICOSLG (from cells)
Fournisseur: ProSci Inc.
91-430
PRSI91-430EA 654 EUR InStock
PRSI91-430
Human recombinant ICOSLG (from cells)
Protéines et Peptides
Inducible Co-Stimulator Ligand (ICOSLG) belongs to the immunoglobulin superfamily. ICOSLG contains Ig-like C2-type (immunoglobulin-like) domain and 1 Ig-like V-type (immunoglobulin-like) domain. ICOSLG acts as a costimulatory signal for T-cell proliferation and cytokine secretion, it also induces B-cell proliferation and differentiation into plasma cells. It could play an important role in mediating local tissue responses to inflammatory conditions, as well as in modulating the secondary immune response by co-stimulating memory T-cell function. ICOSLG is widely expressed lymph nodes, leukocytes and spleen, detected on activated monocytes and dendritic cells.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité