Human recombinant LAMP1 (from cells)
Fournisseur: ProSci Inc.
91-436
PRSI91-436EA 549 EUR InStock
PRSI91-436
Human recombinant LAMP1 (from cells)
Protéines et Peptides
Lysosome-Associated Membrane Glycoprotein 1 (LAMP1) is a single-pass type I membrane protein belonging to the LAMP family. LAMP1 is expressed largely in the endosome-lysosome membranes of cells.It shuttles between lysosomes, endosomes, and the plasma membrane. LAMP1 functions to present carbohydrate ligands to selectins and it has also been implicated in tumor cell metastasis. It has been proposed LAMP1 can be used as a therapeutic agent for certain cancers, as well as a marker for lysosomal storage disorders and degranulation on lymphocytes such as CD8+ and NK cells. Cell surface LAMP1 and LAMP2 have been shown to promote adhesion of human peripheral blood mononuclear cells(PBMC) to vascular endothelium, therefore they are possibly involved in the adhesion of PBMCs to the site of inflammation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité