Human recombinant LAMP2 (from cells)
Fournisseur: ProSci Inc.
91-437
PRSI91-437EA 490 EUR InStock
PRSI91-437
Human recombinant LAMP2 (from cells)
Protéines et Peptides
Lysosomal Associated Membrane Protein 2 (LAMP2) is a major component of lysosomal membranes. LAMP2 is a transmembrane glycoprotein about 110kDa. Mature human LAMP2 consists of a 347 amino acid (aa) intralumenal domain, a 24 aa transmembrane segment, and a 35 aa cytoplasmic tail . The lumenal domain is organized into two heavily N-glycosylated regions. Alternate splicing generates a human LAMP2 isoform (LAMP2B) with a substituted juxtamembrane lumenal region, cytoplasmic tail and transmenmbrane segment.LAMP2 itself can cleavage lysosomal luminal domain and degradation lysosomal. In the help of chaperone HSC73,LAMP2 mediates the lysosomal uptake in complex with cargo proteins and is required for the lysosomal destruction of autophagic vacuoles.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité