Human recombinant LILRB1 (from cells)
Fournisseur: ProSci Inc.
91-438
PRSI91-438EA 471 EUR InStock
PRSI91-438
Human recombinant LILRB1 (from cells)
Protéines et Peptides
The immunoglobulin-like transcript (ILT) family (also named leukocyte Ig-like receptors (LIR) and monocyte/macrophage Ig-like receptors (MIR)) can be activating and inhibitory immunoreceptors. ILTs are expressed on many leukocyte subsets and regulators of immune responses . ILTs share significant homology with killer cell Ig-like receptors (KIR). Except ILT-6, all ILT family members are type I transmembrane proteins having two or four extracellular Ig-like domains . ILT2 is expressed on most monocytes,dendritic cells,and mature B cells. ILT2 is also expressed on small percentages of T-cells and NK cells. ILT2 can prevents cellular activation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité