Human recombinant LYPD3 (from cells)
Fournisseur: ProSci Inc.
91-440
PRSI91-440EA 549 EUR InStock
PRSI91-440
Human recombinant LYPD3 (from cells)
Protéines et Peptides
Ly6/PLAUR domain containing3 (LYPD-3) is a GPI-linked protein. The structure of LYPD-3 is similar to the urokinasetype plasminogen activator receptor (uPAR). LYPD-3 is a 6 -100 kDa molecule with variable cell type-specific N-O-linked glycosylation, mature human LYPD-3 contains two uPAR/Ly6 domains and a Ser/Thr/Pro-rich (STP) region includes a protease sensitive site . The interaction of LYPD-3 with Laminin 1 and 5 on neighboring cells promotes the adhesion, spreading, and migration of tumor cells. LYPD-3 additionally interacts with Galectin-3 and the anterior gradient proteins AG-2 and AG-3. LYPD-3 overexpression in non-small cell lung cancer is predictive of increased mortality.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité