Human recombinant nectin-1 (from cells)
Fournisseur: ProSci Inc.
91-444
PRSI91-444EA 490 EUR InStock
PRSI91-444
Human recombinant nectin-1 (from cells)
Protéines et Peptides
Nectin-1 is a type I transmembrane glycoprotein belonging to the Ig superfamily. Nectin-1 promotes cell-cell contacts by forming homophilic or heterophilic trans-dimers. Heterophilic interactions have been detected between Nectin-1 and Nectin-3 and between Nectin-1 and Nectin-4. Nectin ECDs contain three Ig like domains: an N terminal V type that mediates ligand binding, and two C2 type. Nectin-1 binds viral Glycoprotein D to mediate Herpesvirus (but not Poxvirus) entry into vaginal mucosa, sensory neurons and fibroblasts. In forming adherens junctions and synapses, Nectin-1 and Nectin-3 initiate cell-cell interactions, recruiting alphav beta 3 integrin extracellularly and cadherins intracellularly through afadin and other junctional proteins. These interactions organize the cytoskeleton, strengthen attachment to basement membrane and promote further cell-cell connections. Nectin-1 and Nectin-3 have been found to localize assymetrically along the chemical synapse, with Nectin-1 primarily on the axonal side and Nectin-3 on the dendritic side. Deficiency of Nectin-1 can result in cleft lip/palate ectodermal dysplasia. Nectin-1 downregulation in epithelial cancers is mediated in part by ectodomain shedding, but it may contribute to invasiveness.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité