Human recombinant OSM R beta (from cells)
Fournisseur: ProSci Inc.
91-448
PRSI91-448EA 490 EUR InStock
PRSI91-448
Human recombinant OSM R beta (from cells)
Protéines et Peptides
Oncostatin-M-Specific Receptor Subunit beta (OSMR beta ) is a 150 - 180 kDa member of the IL-6 receptor family. OSMR beta associates with gp130 to form the type II OSM receptor, the receptor is responsive to OSM. Gp130 subunit is shared by other IL-6 family cytokine receptors, and OSMR beta associates with gp130-like receptor (GPL) to form a receptor complex responsive to IL-31. The human OSMR beta cDNA encodes a 979 amino acid (aa) precursor, the precursor includes a 27 aa signal sequence, a 712 aa extracellular domain (ECD), a 22 aatransmembrane segment, and a 218 aa cytoplasmic domain. The ECD contains one partial and one complete hematopoietin domain, an Ig-like domain, and three Fibronectin type-III domains.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité