Human recombinant VEGFD (from cells)
Fournisseur: ProSci Inc.
91-450
PRSI91-450EA 654 EUR InStock
PRSI91-450
Human recombinant VEGFD (from cells)
Protéines et Peptides
Vascular endothelial growth factor D (VEGF-D) is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family. It is highly expressed in lung, heart, small intestine and fetal lung, and at lower levels in skeletal muscle, colon, and pancreas. VEGF-D is growth factor active in angiogenesis, lymphangiogenesis and endothelial cell growth, stimulating their proliferation and migration and also has effects on the permeability of blood vessels. It may function in the formation of the venous and lymphatic vascular systems during embryogenesis, and also in the maintenance of differentiated lymphatic endothelium in adults. It undergoes a complex proteolytic maturation, generating multiple processed forms that bind and activate VEGFR-2 and VEGFR-3 receptors.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité