Human recombinant SEMA5A (from cells)
Fournisseur: ProSci Inc.
91-451
PRSI91-451EA 584 EUR InStock
PRSI91-451
Human recombinant SEMA5A (from cells)
Protéines et Peptides
Semaphorin-5A (SEMA5A) is a member of the Semaphorin family of axon guidance molecules. SEMA5A is a 140 kDa protein. Class 5 Semaphorins are type I transmembrane glycoproteins with an N- terminal Sema domain and multiple juxtamembrane type 1 Thrombospondin (TSP) repeats within their extracellular domains. SEMA5A is expressed in neuroepithelial cells surrounding retinal axons, oligodendrocytes, the base of limb buds, the mesoderm surrounding cranial vessels , and the cardiac atrial septum and endocardial cushions, Human SEMA5A cDNA encodes a signal sequence, a extracellular domain (ECD), a transmembrane sequence and an cytoplasmic portion. SEMA5A mutations have been implicated in the genetic syndrome,cri-du-chat,while some polymorphisms may increase risk for neurodegenerative diseases such as Parkinson. The expression of SEMA5A may be upregulated in metastatic cancer cells and downregulated in autism.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité