Human recombinant ST6GAL1 (from cells)
Fournisseur: ProSci Inc.
91-452
PRSI91-452EA 884 EUR InStock
PRSI91-452
Human recombinant ST6GAL1 (from cells)
Protéines et Peptides
The ST6GAL1 gene encodes beta -Galactosamide alpha-2,6-Sialyltransferase 1. It is a type II membrane protein and is localized to the trans-Golgi network. It catalyzes 2,6-sialylation of Gal beta 1,4-GlcNAc structures on N-glycans. ST6GAL1 is highly expressed in the liver and other tissues. ST6GAL1 deficiency causes abnormalities in B-cell immunoreactivity. The expression and activity of ST6GAL1 are associated with tumor metastasis in breast and colon cancers. The majority of ST6GAL1 in the liver is cleaved and secreted into the serum and may be used as a biomarker for hepatitis diseases.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité