Human recombinant Aminopeptidase P2 (from cells)
Fournisseur: ProSci Inc.
91-460
PRSI91-460EA 789 EUR InStock
PRSI91-460
Human recombinant Aminopeptidase P2 (from cells)
Enzymes
Xaa-Pro aminopeptidase 2 (XPNPEP2) belongs to the peptidase M24B family of metalloproteases. Human XPNPEP2 is widely expressed in various tissues, such as kidney, lung, heart, placenta, liver, small intestine and colon. However, it doesn’t express in brain, skeletal muscle, pancreas, spleen, thymus, prostate, testis and ovary. XPNPEP2 is a Homotrimer which binds 2 manganese ions per subunit. The metalloprotease XPNPEP2 may play a role in the inflammatory process and other reactions produced in response to injury or infection and the metabolism of the vasodilator bradykinin.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité