Human recombinant pentraxin 2 (from cells)
Fournisseur: ProSci Inc.
91-461
PRSI91-461EA 549 EUR InStock
PRSI91-461
Human recombinant pentraxin 2 (from cells)
Protéines et Peptides
Serum Amyoid P Component (SAP) is a monomeric 25 kDa secreted serum glycoprotein that belongs to the pentraxins family. The members of pentaxin superfamily be characterised by calcium dependent ligand binding and distinctive flattened beta -jellyroll structure similar to that of the legume lectins. SAP is a non-fibrillar component, it can interact with DNA and histones. It regulates the solubility of amyloid fibrils and protects them from degradation by proteolytic enzymes and phagocytic cells. SAP scavenge nuclear material released from damaged circulating cells. It has been proposed that SAP may function as an opsonin for a variety of ligands including autoantigens, apoptotic cells, chromatin and micro-organisms.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité