Human recombinant beta-1,4-Galactosyltransferase 4 (from Cells)
Fournisseur: ProSci Inc.
91-464
PRSI91-464EA 654 EUR InStock
PRSI91-464
Human recombinant beta-1,4-Galactosyltransferase 4 (from Cells)
Enzymes
beta -1,4-galactosyltransferase 4 (B4GALT4) is a single-pass type II membrane protein that belongs to the Glycosyltransferase 7 family. B4GALT4 consist of the following 2 domains: N-Acetyllactosamine Synthase and beta -N-Acetylglucosaminyl-Glycolipid beta -1,4-Galactosyltransferase. B4GALT4 is highly expressed in the heart, placenta, kidney, and pancreas; it is lowly expressed in the brain, colon, lung, muscle, ovary, testis, and uterus. B4GALT4 function is responsible for the synthesis of complex-type N-linked oligosaccharides in many glycoproteins as well as the carbohydrate moieties of glycolipids.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité