Human recombinant CD7 (from cells)
Fournisseur: ProSci Inc.
91-467
PRSI91-467EA 654 EUR InStock
PRSI91-467
Human recombinant CD7 (from cells)
Protéines et Peptides
T-Cell Antigen CD7 is a single-pass type I membrane protein that that belongs to the the immunoglobulin superfamily. Human CD7 is synthesized as a 240 amino acid precursor that contains a 25 amino acid signal sequence and a 215 amino acid mature chain with a Ig-like (immunoglobulin-like) domain. CD7 is normally expressed on all T-lymphocytes, NK-cells, pre-B lymphocytes and pleuripotent hematopoietic stem cells. CD7 plays an essential role in T-cell interactions, T-cell/B-cell interaction during early lymphoid development, T- and NK-cell activation and cytokine production. CD7 has been shown to interact with PIK3R1and SECTM1. However, the function of the CD7 protein in the immune system is still largely unknown.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité