Human recombinant Complement Factor H-related 2 (from cells)
Fournisseur: ProSci Inc.
91-468
PRSI91-468EA 654 EUR InStock
PRSI91-468
Human recombinant Complement Factor H-related 2 (from cells)
Protéines et Peptides
Complement Factor H-Related Protein 2 (CFHR2) is a secreted protein that belongs to the complement factor H protein family. Members of the H-related protein family are exclusively composed of individually folded protein domains, termed short consensus repeats (SCRs) or complement control modules. CFHR2 is synthesized as a 270 amino acid precursor that contains an 18 amino acid signal peptide and a 252 amino acid mature chain with 4 Sushi (CCP/SCR) domains. CFHR2 is synthesized in the liver and secreted into plasma. It may be involved in complement regulation. CFHR2 can also be associated with lipoproteins and may play a role in lipid metabolism.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité