Human recombinant FGF R5 (from cells)
Fournisseur: ProSci Inc.
91-471
PRSI91-471EA 528 EUR InStock
PRSI91-471
Human recombinant FGF R5 (from cells)
Protéines et Peptides
Fibroblast Growth Factor Receptor-Like 1 (FGFRL1) is a single-pass type I membrane protein that belongs to the FGF receptor family. The mature human FGFRL1 consists of a 354 amino acid extracellular domain (ECD) with 3 Ig-like C2-type domains, a 21 amino acid transmembrane segment, and a 134 amino acid cytoplasmic domain. FGFR1 expressed in various tissues, preferentially in cartilaginous tissues and pancreas. It highly expressed in the liver, kidney, heart, brain and skeletal muscle, weakly expressed in the lung, small intestine and spleen. FGFRL1 has a negative effect on cell proliferation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité