Human recombinant MINPP1 (from cells)
Fournisseur: ProSci Inc.
91-475
PRSI91-475EA 654 EUR InStock
PRSI91-475
Human recombinant MINPP1 (from cells)
Protéines et Peptides
Multiple Inositol Polyphosphate Phosphatase 1/MINPP1 is an enzyme that removes 3-phosphate from inositol phosphate substrates. MINPP1 also converts 2,3 bisphosphoglycerate (2,3-BPG) to 2-phosphoglycerate. MINPP1 is synthesized as a 487 amino acid precursor that contains an 30 amino acid signal peptide and a 457 amino aicd mature chain. MINPP1 is widely expressed with the highest levels found in kidney, liver and placenta. It acts as a phosphoinositide 5- and phosphoinositide 6-phosphatase and regulates cellular levels of inositol pentakisphosphate (InsP5) and inositol hexakisphosphate (InsP6). MINPP1 may play a role in bone development (endochondral ossification).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité