Human recombinant REG1A (from cells)
Fournisseur: ProSci Inc.
91-480
PRSI91-480EA 584 EUR InStock
PRSI91-480
Human recombinant REG1A (from cells)
Protéines et Peptides
Regenerating Islet-Derived Protein 1- alpha (REG1A) belongs to the Reg family of secreted proteins with a C-type lectic domain. REG1A is highly expressed levels in fetal and infant brains, much lower in adult brains. REG1A promotes the maintenance and growth of pancreatic islet cells and intestinal villi. In addition to, REG1A Might act as an inhibitor of spontaneous calcium carbonate precipitation and be associated with neuronal sprouting in brain, and with brain and pancreas regeneration.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité