Human recombinant serpin A3 (from cells)
Fournisseur: ProSci Inc.
91-484
PRSI91-484EA 852 EUR InStock
PRSI91-484
Human recombinant serpin A3 (from cells)
Protéines et Peptides
Serpin A3 belongs to the Serpin superfamily of serine protease inhibitors. Serpin A3 has been shown to inhibit some serine proteases, such as neutrophil cathepsin G and mast cell chymase. Serpin A3 is synthesized initially in the liver and secreted in plasma. Serpin A3 has been found in the amyloid plaques from the hippocampus of Alzheimer disease brains. In addition to, Serpin A3 is associated with liver disease and Parkinson disease and chronic obstructive pulmonary disease.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité